missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-c-Cbl (Tyr700) Recombinant Rabbit Monoclonal Antibody (CblY700-E1), APC, Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£185.00 - £463.00

Specifications

Antigen Phospho-c-Cbl (Tyr700)
Clone CblY700-E1
Applications Flow Cytometry
Classification Recombinant Monoclonal
Conjugate APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17196938
View Documents
Invitrogen™
MA537353
10 Tests
£185.00
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17136698
GREENER_CHOICE
View Documents
Invitrogen™
MA537107
100 Tests
£463.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
Specifications

Specifications

Phospho-c-Cbl (Tyr700)
Flow Cytometry
APC
Rabbit
Human, Mouse, Rat
P22681, P22682
12402, 500985, 867
A synthetic phospho-peptide corresponding to residues surrounding Tyr700 of human phospho c-Cbl.
Primary
4°C
Cbl
CblY700-E1
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL
Cbl
IgG κ
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.