missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-beta Catenin (Tyr333) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537546
This item is not returnable.
View return policy
Description
A suggested positive control for Western blot is 293 cells; suggested positive control for IHC is human lung carcinoma.
Phospho-beta Catenin (Tyr333) Polyclonal antibody specifically detects Phospho-beta Catenin (Tyr333) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications
| Phospho-beta Catenin (Tyr333) | |
| Polyclonal | |
| Unconjugated | |
| CTNNB1 | |
| armadillo; armadillo homolog; Bcatenin; b-catenin; beta 1 88kDa; beta catenin; Betacatenin; beta-catenin; beta-catenin 1; Bfc; C20orf33; Cadherin associated protein; catenin; catenin (cadherin associated protein), beta 1; catenin (cadherin associated protein), beta 1, 88kDa; catenin (cadherin-associated protein) beta 1; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; Catenin b 1; Catenin b1; catenin beta; catenin beta 1; catenin beta 1 L homeolog; Catenin beta1; Catenin beta-1; Catenin β 1; Catenin β1; Catnb; CHBCAT; CTNB1; CTNNB; ctnnb1; ctnnb1.L; ctnnb1-a; ctnnb1-b; dJ633O20.1; DKFZp686D02253; FLJ25606; FLJ37923; id:ibd2058; Mesc; MRD19; NAP; NYD-SP19; OK/SW-cl.35; OTTHUMP00000209289; P14L; PP8304; PRO2286; RP5-1118M15.1; wu:fb73e10; wu:fi81c06; wu:fk25h01; XELAEV_18031149mg; β catenin; βcatenin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12387, 1499, 84353 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35222, Q02248, Q9WU82 | |
| CTNNB1 | |
| Peptide sequence around phosphorylation site of tyrosine 333 (Y-T-Y(p)-E-K) derived from Human Beta-catenin | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction