missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Bcr (Tyr360) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104787
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Bcr only when phosphorylated at Tyrosine 360.
A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Phospho-Bcr (Tyr360) | |
| Polyclonal | |
| Unconjugated | |
| Bcr | |
| 5133400C09Rik; ABL; AI561783; AI853148; ALL; BCR; BCR activator of RhoGEF and GTPase; BCR protein; BCR, RhoGEF and GTPase activating protein; bcr/abl; BCR/FGFR1 chimera protein; BCR1; breakpoint cluster region; breakpoint cluster region homolog; breakpoint cluster region protein; c-ABL; c-abl oncogene 1; CML; D22S11; D22S662; EC 2.7.11.1; FGFR1/BCR chimera protein; JTK7; Kiaa3017; mKIAA3017; p150; PHL; renal carcinoma antigen NY-REN-26; RP11-83J21.1; v-abl | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 110279, 309696, 613 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11274, Q6PAJ1 | |
| Bcr | |
| A synthesized peptide derived from human BCR(Accession P11274), corresponding to amino acid residues around phosphorylated Tyr360. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction