missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Bcl-6 (Ser333) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539779
This item is not returnable.
View return policy
Description
BCL-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. BCL-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin′s lymphomas and BCL-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt′s lymphomas and in nodular, lymphocyte predomit Hodgkin′s disease.
Specifications
| Phospho-Bcl-6 (Ser333) | |
| Polyclonal | |
| Unconjugated | |
| BCL6 | |
| BCL5, BCL6A, LAZ3, ZBTB27, ZNF51, B-cell lymphoma 5 protein, B-cell lymphoma 6 protein, B-cell lymphoma 6 protein transcript, BCL-5, BCL-6, cys-his2 zinc finger transcription factor, lymphoma-associated zinc finger gene on chromosome 3, protein LAZ-3, zinc finger and BTB domain-containing protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12053, 303836, 604 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41182, P41183 | |
| BCL6 | |
| A synthetic phosphopeptide derived from human Bcl-6 around the phosphorylation site of Ser333 (P-Q-SP-P-Q) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction