missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-BAD (Ser112) Monoclonal Antibody (G.445.9)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA515085
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The protein encoded by BAD is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform.
Specifications
| Phospho-BAD (Ser112) | |
| Monoclonal | |
| 66 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| O35147, Q61337, Q92934 | |
| BAD | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer112 of mouse Bad. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| G.445.9 | |
| Unconjugated | |
| BAD | |
| AI325008; BAD; Bbc2; Bbc6; Bcl2 antagonist of cell death; BCL2 associated agonist of cell death; bcl-2 associated death agonist; Bcl2-antagonist of cell death; BCL2-antagonist of cell death protein; bcl2-associated agonist of cell death; bcl2-associated death promoter; bcl-2-binding component 6; BCL2-binding component 6; BCL2-binding protein; BCL2L8; bcl2-L-8; Bcl-2-like protein 8; BCL-X/BCL-2 binding protein; bcl-XL/Bcl-2-associated death promoter; hypothetical protein LOC615013; OTTMUSP00000017561; OTTMUSP00000022400 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12015, 572, 64639 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction