missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-ATR (Ser428) Recombinant Rabbit Monoclonal Antibody (7F9W10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551545
This item is not returnable.
View return policy
Description
Immunogen sequence: GISPK.
The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
Specifications
| Phospho-ATR (Ser428) | |
| Recombinant Monoclonal | |
| 0.215 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q13535 | |
| ATR | |
| A synthetic phosphorylated peptide around S428 of human ATR (NP_001175.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 7F9W10 | |
| Unconjugated | |
| ATR | |
| 2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 545 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction