missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Ask1 (Thr838) Recombinant Rabbit Monoclonal Antibody (Ask1T838-8D12), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537187
This item is not returnable.
View return policy
Description
MAP3K5 (ASK1) is a serine/threonine kinase that is abundantly expressed in human heart and pancreas and are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activates c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.
Specifications
| Phospho-Ask1 (Thr838) | |
| Recombinant Monoclonal | |
| FITC | |
| MAP3K5 | |
| 7420452D20Rik; apoptosis signal regulating kinase 1; apoptosis signal-regulating kinase 1; ASK; ASK 1; ASK1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; Map3k5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; Mekk5; mitogen activated protein kinase kinase kinase 5; mitogen-activated protein kinase kinase kinase 5; RGD1306565; RP3-325F22.4 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 26408, 4217 | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry | |
| Ask1T838-8D12 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| O35099, Q99683 | |
| MAP3K5 | |
| A synthetic phospho-peptide corresponding to residues surrounding Thr838 of human phospho Ask1. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction