missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ASK1 (Ser83) Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£125.00 - £280.00
Specifications
| Antigen | Phospho-ASK1 (Ser83) |
|---|---|
| Concentration | 220 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18060466
|
Proteintech
28847-1-AP-100UL |
100 μL |
£280.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18015395
|
Proteintech
28847-1-AP-20UL |
20 μL |
£125.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-ASK1 (Ser83) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-ASK1 (Ser83). This antibody recognizes human antigen. The Phospho-ASK1 (Ser83) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-ASK1 (Ser83) | |
| Western Blot, ELISA | |
| Rabbit | |
| 4217 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 220 μg/mL | |
| Liquid | |
| Human | |
| ASK 1, ASK1, MAP3K5, MAPK/ERK kinase kinase 5, MAPKKK5, MEK kinase 5, MEKK 5, MEKK5, Phospho-ASK1 (Ser83) | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title