missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-AS160 (Ser588) Recombinant Rabbit Monoclonal Antibody (5H9L9)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 702305

Produktkod. 15973834

  • £424.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

This antibody is predicted to react with Monkey, Bovine, and Mouse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

AS160 (TBC1D4) is a GTPase-activiting protein for RAB2A, RAB8A, RAB10, and RAB14. The isoform 2 version of this protein promotes the insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane. This movement increases glucose uptake. Mutations associated with the gene cause Diabetes mellitus, non-insulin-dependent, 5 (NIDDM5).
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

Phospho-AS160 (Ser588)
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O60343
TBC1D4
Peptide corresponding to human Phospho-AS160 (Ser588) (aa583-aa592).
100 μg
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
5H9L9
Unconjugated
TBC1D4
5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4
Rabbit
Protein A
RUO
9882
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.