missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AS160 (Ser588) Recombinant Superclonal™ Antibody (5HCLC)
Phospho-AS160 (Ser588) Recombinant Polyclonal Antibody (5HCLC)
Brand: Invitrogen 711337
This item is not returnable.
View return policy
Description
Phospho-AS160 (Ser588) Recombinant Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-AS160 (Ser588) | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O60343 | |
| TBC1D4 | |
| Peptide corresponding to human Phospho-AS160 (Ser588) (aa583-aa592). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 5HCLC | |
| Unconjugated | |
| TBC1D4 | |
| 5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction