missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-AS160 (Ser570) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105405
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of AS160 only when phosphorylated at Ser570.
AS160 (TBC1D4) is a GTPase-activiting protein for RAB2A, RAB8A, RAB10, and RAB14. The isoform 2 version of this protein promotes the insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane. This movement increases glucose uptake. Mutations associated with the gene cause Diabetes mellitus, non-insulin-dependent, 5 (NIDDM5).
Specifications
| Phospho-AS160 (Ser570) | |
| Polyclonal | |
| Unconjugated | |
| TBC1D4 | |
| 5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 210789, 306117, 9882 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60343, Q8BYJ6 | |
| TBC1D4 | |
| A synthesized peptide derived from human TBC1D4(Accession O60343), corresponding to amino acid residues around phosphorylated Ser570. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction