missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-APP (Thr668) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£125.00 - £280.00
Specifications
| Antigen | Phospho-APP (Thr668) |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17521386
|
Proteintech
28788-1-AP-20UL |
20 μL |
£125.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17511386
|
Proteintech
28788-1-AP-100UL |
100 μL |
£280.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-APP (Thr668) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-APP (Thr668). This antibody recognizes human antigen. The Phospho-APP (Thr668) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-APP (Thr668) | |
| Polyclonal | |
| Rabbit | |
| 351 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Human | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title