missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Androgen Receptor (Ser213, Ser210) Monoclonal Antibody (156C135.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516221
This item is not returnable.
View return policy
Description
Suggested positive control: IGF-1 stimulated LNCaP. Predicted cross-reactivity with Chimpanzee, Canine, Mouse, Porcine and Rat. Immunogen sequence: GRAREASGAP TSSKD.
The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis.AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain.Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy's disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).
Specifications
| Phospho-Androgen Receptor (Ser213, Ser210) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P10275, P15207, P19091, Q9TT90 | |
| AR | |
| Synthetic peptide corresponding to amino acids 207-221 of human androgen receptor, containing the serine 213 phosphorylation site. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 156C135.2 | |
| Unconjugated | |
| AR | |
| AIS; Andr; androgen nuclear receptor variant 2; androgen receptor; androgen receptor (Testicular feminization), same as Tfm; androgen receptor splice variant 4b; Ar; AR8; AR-A; AW320017; DHTR; dihydrotestosterone receptor; HUMARA; HYSP1; KD; NR3C4; Nuclear receptor subfamily 3 group C member 4; RP11-383C12.1; SBMA; SMAX1; TFM; Variant AR45 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11835, 24208, 367, 403588 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction