missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-AMPK alpha-1,2 (Thr172) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517831
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.
Specifications
| Phospho-AMPK alpha-1,2 (Thr172) | |
| Polyclonal | |
| Unconjugated | |
| PRKAA2 | |
| 2310008I11Rik; 5'-AMP-activated protein kinase alpha-1 catalytic subunit; 5'-AMP-activated protein kinase alpha-2 catalytic subunit; 5'-AMP-activated protein kinase catalytic alpha-2 subunit; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; 63 kDa subunit; A830082D05; AAPK2; ACACA kinase; acetyl-CoA carboxylase kinase; AI194361; AI450832; AL024255; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase; AMP-activated protein kinase alpha 2; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK; AMPK 63 kDa subunit; AMPK alpha 1; AMPK alpha 2; AMPK alpha-1 chain; AMPK alpha-2 chain; AMPK subunit alpha-1; AMPK subunit alpha-2; AMPK1; AMPK2; AMPKa1; AMPKa2; AMPK-activated protein kinase alpha-2 subunit; AMPKalpha1; AMPK-alpha-1 chain; AMPKalpha2; AMPK-alpha-2 chain; C130083N04Rik; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; pAMPK; PRKAA; Prkaa1; Prkaa2; protein kinase; protein kinase AMP-activated alpha 2 catalytic subunit; protein kinase AMP-activated catalytic subunit alpha 1; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 1 catalytic subunit; protein kinase, AMP-activated, alpha 2 catalytic subunit; tau-protein kinase PRKAA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 105787, 108079, 5562, 5563, 65248, 78975 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 83 μg/mL | |
| 0.01M HEPES with 0.15M NaCl, 50% glycerol, 100μg/mL BSA and no preservative; pH 7.5 | |
| P54645, P54646, Q09137, Q13131, Q5EG47, Q8BRK8 | |
| PRKAA1, PRKAA2 | |
| Synthetic phosphopeptide corresponding to residues surrounding Thr172 of human AMPKa. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction