missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-AMPK alpha-1,2 (Thr183, Thr172) Polyclonal Antibody

Description
A suggested positive control for Western blot is C2C12 cells; suggested positive control for IHC is human lung carcinoma.
The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.
Specifications
Specifications
| Antigen | Phospho-AMPK alpha-1,2 (Thr183, Thr172) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PRKAA2 |
| Gene Accession No. | P54645, P54646, Q09137, Q13131, Q5EG47, Q8BRK8 |
| Gene Alias | 2310008I11Rik; 5'-AMP-activated protein kinase alpha-1 catalytic subunit; 5'-AMP-activated protein kinase alpha-2 catalytic subunit; 5'-AMP-activated protein kinase catalytic alpha-2 subunit; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; 63 kDa subunit; A830082D05; AAPK2; ACACA kinase; acetyl-CoA carboxylase kinase; AI194361; AI450832; AL024255; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase; AMP-activated protein kinase alpha 2; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK; AMPK 63 kDa subunit; AMPK alpha 1; AMPK alpha 2; AMPK alpha-1 chain; AMPK alpha-2 chain; AMPK subunit alpha-1; AMPK subunit alpha-2; AMPK1; AMPK2; AMPKa1; AMPKa2; AMPK-activated protein kinase alpha-2 subunit; AMPKalpha1; AMPK-alpha-1 chain; AMPKalpha2; AMPK-alpha-2 chain; C130083N04Rik; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; pAMPK; PRKAA; Prkaa1; Prkaa2; protein kinase; protein kinase AMP-activated alpha 2 catalytic subunit; protein kinase AMP-activated catalytic subunit alpha 1; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 1 catalytic subunit; protein kinase, AMP-activated, alpha 2 catalytic subunit; tau-protein kinase PRKAA1 |
| Gene Symbols | PRKAA1, PRKAA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?