missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-alpha Catenin (Ser641) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538750
This item is not returnable.
View return policy
Description
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
Specifications
| Phospho-alpha Catenin (Ser641) | |
| Polyclonal | |
| Unconjugated | |
| CTNNA1 | |
| Catenin alpha-1, Alpha E-catenin, Cadherin-associated protein, Renal carcinoma antigen NY-REN-13, CTNNA1, Catenin-alpha1, Catenin alpha1, alphaE-catenin, Catenin alpha-2, Alpha N-catenin, Alpha-catenin-related protein, CTNNA2, CAPR, Catenin-alpha2, Catenin alpha2, alphaN-catenin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12385, 1495, 307505 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35221, P26231 | |
| CTNNA1 | |
| A synthetic phosphopeptide derived from human Catenin-alpha1 around the phosphorylation site of Ser641 (D-D-SP-D-F) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction