missing translation for 'onlineSavingsMsg'
Learn More

Phospho-alpha Adducin (Ser726) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Product Code. 16604375
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16604375 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16604375 Supplier Invitrogen Supplier No. PA5104878

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-alpha Adducin (Ser726) Polyclonal antibody specifically detects Phospho-alpha Adducin (Ser726) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-alpha Adducin (Ser726)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADD1
Gene Accession No. Q9QYC0, P35611, Q63028
Gene Alias ADD1; ADD2; ADDA; ADDB; adducin 1; adducin 1 (alpha); Adducin beta; Adducin1; Adducin-1; Adducin-alpha; Adducin-beta; AI256389; Alpha adducin; alpha-ADD; Alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; Erythrocyte adducin subunit alpha
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADD1 around the phosphorylation site of Ser726.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118, 11518, 24170
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.