Learn More
Phospho-AKT (Ser473) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28731-1-AP-20UL
Description
The Phospho-AKT (Ser473) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-AKT (Ser473). This antibody recognizes human, mouse, rat antigen. The Phospho-AKT (Ser473) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-AKT (Ser473) | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| 20 μL | |
| 207 | |
| IgG |