missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Afadin (Tyr1230) Polyclonal Antibody

Phospho-Afadin (Tyr1230) Polyclonal Antibody
Brand: Invitrogen PA5105593
This item is not returnable.
View return policy
Description
Phospho-Afadin (Tyr1230) Polyclonal Antibody for Western Blot
Specifications
| Phospho-Afadin (Tyr1230) | |
| Polyclonal | |
| Unconjugated | |
| Afdn | |
| 5033403D15Rik; Af6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; afadin; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; MLLT4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein Af-6; RP3-431P23.3; RP3-470B24.4; S-afadin | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 17356, 26955, 4301 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35889, P55196, Q9QZQ1 | |
| Afdn | |
| A synthesized peptide derived from human AFDN(Accession P55196), corresponding to amino acid residues around phosphorylated Tyr1230. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction