missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-4EBP1 (Thr70) Recombinant Rabbit Monoclonal Antibody (5N1L9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551551
This item is not returnable.
View return policy
Description
Phospho-4EBP1 (Thr70) Recombinant Monoclonal Antibody for Western Blot, ELISA
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
Specifications
| Phospho-4EBP1 (Thr70) | |
| Recombinant Monoclonal | |
| 0.56 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q13541, Q60876 | |
| EIF4EBP1 | |
| A synthetic phosphorylated peptide around T70 of human eIF4EBP1 (NP_004086.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 5N1L9 | |
| Unconjugated | |
| EIF4EBP1 | |
| 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13685, 1978 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction