missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-4EBP1 (Ser65) Recombinant Rabbit Monoclonal Antibody (9M4D7)
Description
Immunogen Sequence: RNSPV.
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
Specifications
Specifications
| Antigen | Phospho-4EBP1 (Ser65) |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 9M4D7 |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 |
| Gene | EIF4EBP1 |
| Gene Accession No. | Q13541, Q60876 |
| Gene Alias | 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?