missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Phosphatesmo MI Qualitative Test Paper

Product Code. 10144761 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
White to Yellow
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10144761 White to Yellow Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144761 Supplier Macherey-Nagel™ Supplier No. 90612

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Macherey-Nagel Phosphatesmo MI Qualitative Test Paper, Color Indicator: White to yellow, For Use With: For detection of Alkaline Phosphatase in milk, Quantity: 50Pack, Storage Requirements: 2 to 8 deg.C

Specifications

Color Indicator White to Yellow
For Use With (Application) For detection of Alkaline Phosphatase in milk
Quantity 50 Pack
Storage Requirements 2°C to 8°C
Type Qualitative Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.