missing translation for 'onlineSavingsMsg'
Learn More

Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL

Product Code. 11475010
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475010 500 mL 500mL
11363647 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475010 Supplier Thermo Scientific Alfa Aesar Supplier No. 035548.AP
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol

Phosphate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym Phosphate, Ion chromatography standard solution
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium;dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Physical Form Liquid
Concentration PO4-3 1000 μg/mL
Solubility Miscible with water
Quantity 500 mL
Chemical Name or Material Phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.