missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), with Triton∣r X-100

Product Code. 15400317
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15400317

Brand: Thermo Scientific Chemicals J63521.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for maintaining a constant pH.

Phosphate-buffered saline is used in biological research. PBS buffer is ideal for maintaining a constant pH, and since it is isotonic to cells. PBS is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption. And also used in immunoassay procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
Physical Form Liquid
Quantity 250 mL
compliance-icons
Product Identifier
  • Phosphate-buffered saline (PBS, 10X), with Triton X-100
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Phosphoric acid, disodium salt

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt