missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula

Product Code. 15440217
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Unit Size:
1 L
2 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440217 500 mL 500mL
15450217 1 L 1 L
15460217 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440217 Supplier Thermo Scientific Chemicals Supplier No. J61917.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells.

Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula is used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells. It plays an important role in the transportation of tissue samples and as diluents in various reagent preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 500 mL
Packaging Plastic bottle
What are the concentrations of the components in Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula?

Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula contains 89 mM sodium phosphate dibasic, 15 mM potassium phosphate monobasic, 1.37 mM sodium chloride, and 27 mM potassium chloride.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.