missing translation for 'onlineSavingsMsg'
Learn More

PBS, Phosphate Buffered Saline, 10X Solution, pH 7.4, CellPURE™, Fisher BioReagents

Product Code. 10212990
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10212990 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10212990 Supplier Fisher BioReagents Supplier No. BP29404

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Usage: When diluted to a 1X concentration using water, this product yields 0.01M phosphate buffer and 0.154M NaCl, pH 7.4 at 25°C

  • CellPURE Phosphate Buffered Saline provides an optimal formulation for cell biology experiments in which the osmolarity of cells must be maintained.
  • PBS can also be used in biochemistry studies for maintaining proteins in a certain pH range.
  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
  • Tested for the absence of DNase, RNase, nickase, and protease
  • Endotoxin, cell culture and bioburden testing -- this PBS solution is suitable for cell culture applications
  • This 10X concentrate will yield a 1X PBS solution containing 0.01M phosphate, 0.154M NaCl, and a pH of 7.4 at 25°C
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 4 L
Packaging PolyPac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.