missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer Solution, pH 7, Honeywell™ Fluka™

Product Code. 15656930 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly bottle
Poly pail
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15656930 1 L Poly bottle 1 L
15636930 10 L Poly pail 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15656930 Supplier Honeywell Fluka Supplier No. 731731L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5

Specifications

Density 1.01g/mL at 20°C
Quantity 1 L
Packaging Poly bottle
UN Number NONH for all modes of transport
Chemical Name or Material Phosphate buffer solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.