missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate buffer solution, pH 6 EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
| CAS | 7732-18-5 |
| pH | 6 |
| Quantity | 1000 mL |
| Packaging | Bottle |
| Grade | Eu. Ph. Reference |
| Chemical Name or Material | Phosphate buffer solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction