missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate buffer solution, pH 5.5 EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
CAS | 7732-18-5 |
pH | 5.5 |
Quantity | 1000 mL |
Packaging | Bottle |
Grade | Eu. Ph. Reference |
Chemical Name or Material | Phosphate buffer solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'