missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.2

Product Code. 15495189
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495189 1 L 1 L
15485189 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495189 Supplier Thermo Scientific Chemicals Supplier No. J62899.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7558-80-7119.976 g/mol

Sodium phosphate monobasic is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is also used in HPLC. Phosphates are often used in foods and in water treatment. It is added in animal feed, toothpaste, and evaporated milk. It is used as a thickening agent and emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifications

Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 1 L
Packaging Plastic Bottle

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.