missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Phosphatase Inhibitor Cocktail IV
Description
- Specificity: Alkaline Phosphatases and Ser/Thr Phosphatases 1 and 2A (PP-1 and PP-2A)
- Dilute 1:50 before using
Specifications
Specifications
| Quantity | 1 Vial |
| Composition | (-)-p-Bromotetramisole oxalate, Cantharidin, Calyculin A, Discodermia calyx |
Safety and Handling
Product Identifier
- Phosphatase Inhibitor Cocktail
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: L-p-Bromotetramisoleoxalate; 6-Bromolevamisole oxalate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?