missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Phosphatase Inhibitor Cocktail III

Product Code. 12841650
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12841650 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12841650 Supplier Fisher BioReagents™ Supplier No. 12841650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reconstitute with 1mlL water to produce a 1 mL concentrate.isher Bioreagents Phosphatase Inhibitor Cocktail III is a lyophilized blend of four phosphatase inhibitors for use with animal tissue extracts.

  • Specificity: Ser/Thr Phosphatases and Protein Tyrosine Phosphatases
  • Dilute 1:5 before using

Specifications

Product Type Phosphatase Inhibitor Cocktail
Quantity 1 Vial
Composition Sodium Fluoride, Sodium Orthovanadate, Sodium Phyrophosphate, decahydrate B-Glycerophosphate
compliance-icons
Product Identifier
  • Phosphatase Inhibitor Cocktail III
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Pyrophosphate, beta-Glycerophosphate, Sodium Fluoride, Sodium Orthovanadate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.