missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphatase Inhibitor Cocktail II

Product Code. 15443689
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443689 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15443689 Supplier Thermo Scientific Chemicals Supplier No. J61022.AA
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts. This product has been optimized and tested for tyrosine protein phosphatases, acid and alkaline phosphatases. It has been tested on cell extracts from animal tissues including bovine liver and human placenta, and cytosolic extract of rabbit muscle. It is a featured product for Kinase Phosphatase Biology research.An inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases. It is used with tissue and cell extracts, including extracts containing detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100X
Color Colorless
Description Inhibit broad spectrum of serine/theorine and tyrosine phosphatases
Product Type Phosphatase Inhibitor Cocktail
Content And Storage -30°C to -10°C
Form Liquid
Quantity 5 mL
Composition 100mM sodium fluoride, 50mM sodium orthovanadate, 50mM sodium pyrophosphate, and 100mM b-glycerophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.