missing translation for 'onlineSavingsMsg'
Learn More

Phosmet-oxon, TRC

Product Code. 30424360
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424360 10 mg
30418796 50 mg
30416245 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424360 Supplier Toronto Research Chemicals Supplier No. TRCP35370510MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3735-33-9
Molecular Formula C11 H12 N O5 P S
Molecular Weight (g/mol) 301.26
Synonym Phosphorothioic acid, O,O-dimethyl S-phthalimidomethyl ester (6CI), Phosphorothioic acid, O,O-dimethyl ester, S-ester with N-(mercaptomethyl)phthalimide (7CI,8CI), Phthalimide, N-(mercaptomethyl)-, S-ester with O,O-dimethyl phosphorothioate (8CI), Imidoxon, N-(Mercaptomethyl)phthalimide S-(O,O-dimethyl phosphorothioate), O,O-Dimethyl S-phthalimidomethyl phosphorothioate, O,O-Dimethyl phosphorothioate S-ester with N-(mercaptomethyl)phthalimide, Oxoimidan, Phosmet oxon, R 1571, R 1571A, S-Phthalimidomethyl O,O-dimethyl phosphorothioate, Stauffer R 1571, Stauffer R 1571A
IUPAC Name 2-(dimethoxyphosphorylsulfanylmethyl)isoindole-1,3-dione
SMILES COP(=O)(OC)SCN1C(=O)c2ccccc2C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12NO5PS/c1-16-18(15,17-2)19-7-12-10(13)8-5-3-4-6-9(8)11(12)14/h3-6H,7H2,1-2H3
Formula Weight 301.0174
Chemical Name or Material Phosmetoxon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.