missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosducin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93747-0.1ml
This item is not returnable.
View return policy
Description
Phosducin Polyclonal antibody specifically detects Phosducin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Phosducin | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| 33 kDa phototransducing protein, 33kDA phototransducing protein, G beta gamma binding protein, PHD, PhLP, phosducin, phosducin-like orphan protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-246 of human PDC (NP_002588.3). MEEAKSQSLEEDFEGQATHTGPKGVINDWRKFKLESQDSDSIPPSKKEILRQMSSPQSRNGKDSKERVSRKMSIQEYELIHKEKEDENCLRKYRRQCMQDMHQKLSFGPRYGFVYELETGKQFLETIEKELKITTIVVHIYEDGIKGCDALNSSLTCLAAEYPIVKFCKIKASNTGAGDRFSLDVLPTLLIYKGGELISNFISVAEQFAEEFFAGDVESFLNEYGLLPEREVHVLEHTKIEEEDVE | |
| 0.1 mL | |
| Signal Transduction | |
| 5132 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction