missing translation for 'onlineSavingsMsg'
Learn More

Phorbol, MedChemExpress

Product Code. p-200099297
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443946

Brand: Medchem Express HYN2147

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Phorbol is a highly toxic diterpene, whose esters have important biological properties.

Phorbol is a highly toxic diterpene, whose esters have important biological properties.

Specifications

Chemical Name or Material Phorbol
CAS 17673-25-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H28O6
Quantity 25 mg
Synonym 4β-Phorbol
Solubility Information DMSO : 50 mg/mL (137.20 mM; Need ultrasonic) ∣H2O : ≥ 20 mg/mL (54.88 mM)
SMILES O=C1C(C)=C[C@@]2([H])[C@]3(O)[C@@]([C@@](C4(C)C)([H])[C@@]4(O)[C@H](O)[C@H]3C)([H])C=C(CO)C[C@]12O
Molecular Weight (g/mol) 364.43
Formula Weight 364.43
Percent Purity 97.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.