missing translation for 'onlineSavingsMsg'
Learn More

Phorbol, MedChemExpress

Product Code. 16429268
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16429268 10 mg 10mg
16443946 25 mg 25mg
16422589 1 mg 1mg
16411688 5 mg 5mg
16404937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16429268 Supplier Medchem Express Supplier No. HYN2147
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phorbol is a highly toxic diterpene, whose esters have important biological properties.

Phorbol is a highly toxic diterpene, whose esters have important biological properties.

Specifications

Chemical Name or Material Phorbol
CAS 17673-25-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H28O6
Quantity 10 mg
Synonym 4β-Phorbol
Solubility Information DMSO : 50 mg/mL (137.20 mM; Need ultrasonic) ∣H2O : ≥ 20 mg/mL (54.88 mM)
SMILES O=C1C(C)=C[C@@]2([H])[C@]3(O)[C@@]([C@@](C4(C)C)([H])[C@@]4(O)[C@H](O)[C@H]3C)([H])C=C(CO)C[C@]12O
Molecular Weight (g/mol) 364.43
Formula Weight 364.43
Percent Purity 97.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.