missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phorbol 12-myristate 13-acetate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Description
Phorbol 12-myristate 13-acetate (PMA), a phorbol ester, is a dual SphK and protein kinase C (PKC) activator. Phorbol 12-myristate 13-acetate is a NF-κB activator. Phorbol 12-myristate 13-acetate induces differentiation in THP-1 cells.
Specifications
Specifications
| Chemical Name or Material | Phorbol 12-myristate 13-acetate |
| CAS | 16561-29-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H56O8 |
| Quantity | 25 mg |
| Synonym | PMA TPA Phorbol myristate acetate |
| Solubility Information | DMSO : 100 mg/mL (162.12 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (162.12 mM; Need ultrasonic) |
| SMILES | CCCCCCCCCCCCCC(O[C@H]([C@H]1C)[C@]2(OC(C)=O)[C@@]([C@@](C=C(CO)C[C@]34O)([H])[C@@]1(O)[C@]4([H])C=C(C)C3=O)([H])C2(C)C)=O |
| Molecular Weight (g/mol) | 616.83 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction