missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12-myristate 13-acetate, 95%

Product Code. 15496069
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496069 25 mg 25mg
15476069 1 mg 1mg
15466069 5 mg 5mg
15486069 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15496069 Supplier Thermo Scientific Alfa Aesar Supplier No. J63916.MB
  • ONLINE EXCLUSIVE PRICE
    £792.00
    £712.65 / 25mg
    Save £79.35
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Activates protein kinase C and promotes skin tumors in mice | CAS: 16561-29-8 | C36H56O8 | 616.84 g/mol

PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

Solubility
Very soluble in methanol, soluble in DMSO, chloroform, ethanol or acetone, ether, ethyl acetate, DMF or acetonitrile. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16561-29-8
Molecular Formula C36H56O8
Molecular Weight (g/mol) 616.84
MDL Number MFCD00036736
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
PubChem CID 27924
ChEBI CHEBI:37537
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O

Specifications

Color White
Quantity 25 mg
Beilstein 2407201
Sensitivity Light sensitive
Merck Index 14,7332
Solubility Information Very soluble in methanol,soluble in DMSO,chloroform,ethanol or acetone,ether,ethyl acetate,DMF or acetonitrile. Insoluble in water.
Formula Weight 616.83
Percent Purity >94%
Physical Form Film or Powder
Chemical Name or Material Phorbol 12-myristate 13-acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.