missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Phorbol 12-myristate 13-acetate

Product Code. 17160312
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17160312 1 mg 1mg
17170312 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17160312 Supplier STEMCELL Technologies Supplier No. 74042
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PKC pathway activator; Activates protein kinase C

  • Phorbol 12-myristate 13-acetate (PMA), a phorbol ester, is a reversible, highly potent activator of protein kinase C (PKC), including Group A and Group B isoforms (Blumberg)
  • PMA activates PKC isoforms through the binding of the C1 domain (Hurley et al.) PMA can also activate certain mitogen-activated protein (MAP) kinase pathways through PKC (Adams & Parker)
  • Stimulates cardiac differentiation of mesenchymal stem cells (Seo et al.)
  • Promotes hematopoietic differentiation (Clemens et al.)
  • Stimulates differentiation of IL-15-stimulated lamina propria CD4+ T cells into CXCR5+CD4+ cells when used together with ionomicin (Sarra et al.)
  • In combination with AS101, it shows anti-leukemic activity and stimulates human leukemia cell lines to differentiate into macrophage-like cells (Glesne & Huberman; Hayun et al.)

Specifications

CAS 16561-29-8
Synonym 12-O-Tetradecanoylphorbol-13-acetate; PMA; TPA
Quantity 1 mg
Molecular Formula C36H56O8
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.