missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12,13-dibutyrate, MedChemExpress

Product Code. 16460138
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460138 5 mg 5mg
16447678 10 mg 10mg
16441759 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16460138 Supplier Medchem Express Supplier No. HY18985
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phorbol 12,13-dibutyrate (Phorbol dibutyrate) is a PKC activator and a potent skin tumor promoter.

Phorbol 12,13-dibutyrate (Phorbol dibutyrate) is a PKC activator and a potent skin tumor promoter.

Specifications

Chemical Name or Material Phorbol 12,13-dibutyrate
CAS 37558-16-0
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C28H40O8
Quantity 5 mg
Synonym Phorbol dibutyrate PDBu
Solubility Information DMSO : ≥ 125 mg/mL (247.72 mM)
SMILES O=C1C(C)=C[C@@]2([H])[C@]3(O)[C@@]([C@@](C4(C)C)([H])[C@@]4(OC(CCC)=O)[C@H](OC(CCC)=O)[C@H]3C)([H])C=C(CO)C[C@]12O
Molecular Weight (g/mol) 504.61
Formula Weight 504.61
Percent Purity 97.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.