missing translation for 'onlineSavingsMsg'
Learn More

Phorate Oxon Sulfoxide, TRC

Product Code. 18091920
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18091920 10 mg 10mg
18071640 100 mg 100mg
18080040 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18091920 Supplier Toronto Research Chemicals Supplier No. TRCP35352010MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2588-05-8
Molecular Formula C7 H17 O4 P S2
Molecular Weight (g/mol) 260.31
Synonym Phorate oxon sulfoxide, O,O-Diethyl S-(ethylsulfinyl)methyl phosphorothioate
IUPAC Name 1-[ethoxy(ethylsulfinylmethylsulfanyl)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)SCS(=O)CC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H17O4PS2/c1-4-10-12(8,11-5-2)13-7-14(9)6-3/h4-7H2,1-3H3
Formula Weight 260.0306
Percent Purity >95%
Chemical Name or Material Phorate Oxon Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.