missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ PHMT Thermoshaker for Microtubes and Microplates

Accelerates in 3 seconds for fast start but also operates smoothly and quietly, this Grant Instruments™ PMHT Thermoshaker for Microtubes and Microplates is ideal for DNA extraction, enzyme reactions and cellular biology techniques.

Brand:  Grant Instruments™ PHMT-PSC32

Code : 57

Additional Details : Weight : 4.00000kg

 View more versions of this product

Product Code. 15291166

  • £1600.60 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket

Description

Description

  • Combines three instruments in one:
    • Microtube and microplate thermoshaker
    • Compact benchtop incubator
    • Microtube and microplate shaker in cold or ambient temperatures
  • 2-line LCD display clearly indicates both set and actual values for temperature, shaking speed and time.
  • Interchangeable block for 20 or 12 microtubes provides flexibility for an easy change in application.
  • Temperature calibration function to compensate for differences in the thermal behaviour of different tubes and samples.
  • Operates extremely smoothly, quietly and consistently.
  • Includes ‘soft’ start function for delicate samples.
  • Fast start - acceleration 3 seconds.
  • Compact and sturdy, with a low profile and small footprint.
Specifications

Specifications

2 mm
Ambient +5° to 100°C
0.2 mL and 1.5 mL
0.1°C at 37°C; 0.2°C at 60°C; 0.2°C at 100°C
130 mm
4 kg
2 line x 16 character LCD
250 to 1400 rpm (increments 10 rpm)
32 x 0.2 mL and 12 x 1.5 mL Microtubes
5°C to 100°C
230 mm
205 mm
PHMT-PSC32
32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers