missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHLPP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550475
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PHLPP1 protein.
PHLPP1 is a member of the serine/threonine phosphatase family, which are important regulators of Akt serine-threonine kinases (AKT1, AKT2, AKT3) and conventional/novel protein kinase C (PKC) isoforms. PHLPP1 and PHLPP2 have a similar domain structure and have been shown to dephosphorylate and inactivate, distinct Akt isoforms, at one of the two critical phosphorylation sites required for activation: Serine473. PHLPP2 dephosphorylates AKT1 and AKT3, whereas PHLPP1 is specific for AKT2 and AKT3. PHLPP1 promotes apoptosis and may act as a tumor suppressor. Increased expression of PHLPP1 may also play a role in obesity and type 2 diabetes by interfering with Akt-mediated insulin signaling.
Specifications
| PHLPP1 | |
| Polyclonal | |
| Unconjugated | |
| PHLPP1 | |
| AI836256; Circadian Oscillatory Protein (SCOP); EC 3.1.3.16; hSCOP; KIAA0606; mKIAA0606; PH domain and leucine rich repeat protein phosphatase 1; PH domain leucine-rich repeat protein phosphatase 1; PH domain leucine-rich repeat-containing protein phosphatase 1; PH domain-containing family E member 1; Phlpp; PHLPP1; pleckstrin homology domain containing, family E (with leucine rich repeats) member 1; pleckstrin homology domain-containing family E member 1; pleckstrin homology domain-containing family E protein 1; Plekhe1; PPM3A; protein phosphatase, Mg2+/Mn2+ dependent 3A; SCN circadian oscillatory protein; SCOP; Suprachiasmatic nucleus circadian oscillatory protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23239 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O60346 | |
| PHLPP1 | |
| Fusion protein corresponding to residues near the C terminal of human PH domain and leucine rich repeat protein phosphatase 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction