missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHLPP Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81766F
This item is not returnable.
View return policy
Description
PHLPP Polyclonal antibody specifically detects PHLPP in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| PHLPP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunofluorescence | |
| FITC | |
| EC 3.1.3.16, hSCOP, KIAA0606, MGC161555, PH domain and leucine rich repeat protein phosphatase, PH domain and leucine rich repeat protein phosphatase 1, PH domain leucine-rich repeat-containing protein phosphatase 1, PHLPP, pleckstrin homology domain containing, family E (with leucine rich repeats)member 1, Pleckstrin homology domain-containing family E member 1, PLEKHE1, SCN circadian oscillatory protein, SCOPPH domain-containing family E member 1, Suprachiasmatic nucleus circadian oscillatory protein | |
| PHLPP antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PHLPP. The immunogen is located within amino acids 250 - 300 of PHLPP. Amino Acid Squence: REPAEPPPEAGPRLAPPE | |
| 0.1 mL | |
| Cancer, Protein Phosphatase, Tumor Suppressors | |
| 23239 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction