missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHLP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521817
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299. Predicted reactivity: Mouse (90%), Rat (90%), Rhesus Monkey (97%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosducin-like protein is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin, a phosphoprotein expressed in retina and pineal gland. Both phosducin-like protein and phosphoducin have been shown to regulate G-protein signaling by binding to the beta-gamma subunits of G proteins.
Specifications
| PHLP | |
| Polyclonal | |
| Unconjugated | |
| Pdcl | |
| 1200011E13Rik; AW108059; PDCL; PHLOP1; PHLP; PhLP1; phosducin like; phosducin-like; phosducin-like protein; RP11-465F21.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5082, 67466 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q13371, Q9DBX2 | |
| Pdcl | |
| Recombinant protein encompassing a sequence within the center region of human PHLP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction