missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHLP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554480
This item is not returnable.
View return policy
Description
Immunogen sequence: EQCREMERLI KKLSMTCRSH LDEEEEQQKQ KDLQEKISGK MTLKEFAIMN EDQDDEEFLQ QYRKQRMEEM RQQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
Phosducin-like protein is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin, a phosphoprotein expressed in retina and pineal gland. Both phosducin-like protein and phosphoducin have been shown to regulate G-protein signaling by binding to the beta-gamma subunits of G proteins.
Specifications
| PHLP | |
| Polyclonal | |
| Unconjugated | |
| Pdcl | |
| 1200011E13Rik; AW108059; PDCL; PHLOP1; PHLP; PhLP1; phosducin like; phosducin-like; phosducin-like protein; RP11-465F21.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13371 | |
| Pdcl | |
| Recombinant protein corresponding to Human PHLP. Recombinant protein control fragment (Product #RP-92978). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction