missing translation for 'onlineSavingsMsg'
Learn More

Phloxine B, MP Biomedicals™

Product Code. 11477992 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477992 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477992 Supplier MP Biomedicals Supplier No. 0215264625
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18472-87-2
Molecular Formula C20H4Br4Cl4O5Na2
Synonym C.I. 45410, Acid red 92
Color Brown to Red
Quantity 25 g
Physical Form Powder
Formula Weight 829.66
Chemical Name or Material Phloxine B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.