missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phloxine B, 85%, pure, high purity, Biological stain

Product Code. 10501641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10501641 25 g Glass bottle 25g
10133730 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10501641 Supplier Thermo Scientific Chemicals Supplier No. 189470250
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18472-87-2 | C20H2Br4Cl4Na2O5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20H2Br4Cl4Na2O5
MDL Number MFCD00005061
Synonym Acid Red 92, C.I. 45410

Specifications

Absorbance 875 to 1300 (at lambda max.)
Absorption Ratio 1.38 to 2.24
Dye Content 80% min. (on anhydrous substance)
Infrared Spectrum Authentic
Lambda Max 546 to 550nm (in 50:50 METHS:water)
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Assay Percent Range 85%
Beilstein 19, IV, 2921
Merck Index 15, 7440
Color Brown to Brown/Red
Quantity 25 g
Physical Form Powder
Formula Weight 829.64
Percent Purity 85%
Grade Pure
Chemical Name or Material Phloxine B
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.