missing translation for 'onlineSavingsMsg'
Learn More

Phloretin, TRC

Product Code. 30137957
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137957 1 g 1g
30007314 100 mg 100mg
30004607 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137957 Supplier Toronto Research Chemicals Supplier No. TRCP3390001G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-82-2
Molecular Formula C15 H14 O5
Molecular Weight (g/mol) 274.27
Synonym 1-Propanone, 3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)-, Phloretin (6CI), Propiophenone, 2',4',6'-trihydroxy-3-(p-hydroxyphenyl)- (8CI), 2',4',6'-Trihydroxy-3-(4-Hydroxyphenyl)propiophenone, 2',4',6'-Trihydroxy-3-(p-hydroxyphenyl)propiophenone, 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)-1-propanone, Dihydronaringenin, NSC 407292, Naringenin dihydrochalcone, Phloretol, RJC 02792, β-(p-Hydroxyphenyl)-2,4,6-trihydroxypropiophenone, β-(p-Hydroxyphenyl)phloropropiophenone
IUPAC Name 3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
SMILES Oc1ccc(CCC(=O)c2c(O)cc(O)cc2O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H14O5/c16-10-4-1-9(2-5-10)3-6-12(18)15-13(19)7-11(17)8-14(15)20/h1-2,4-5,7-8,16-17,19-20H,3,6H2
Formula Weight 274.0841
Chemical Name or Material Phloretin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.